Merck
  • Quantitation of quinapril in human plasma by matrix-assisted laser desorption ionization time-of-flight mass spectrometry with quinolone matrix additives.
Quantitation of quinapril in human plasma by matrix-assisted laser desorption ionization time-of-flight mass spectrometry with quinolone matrix additives.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-08-27)
Chi-Yu Lu, Fei-Tsui Liu, Chia-Hsien Feng
ABSTRACT
The renin-angiotensin-aldosterone system (RAAS) is an essential body fluid maintenance system that controls pressure in the human body. The conversion of angiotensin I to angiotensin II by angiotensin-converting enzyme (ACE) is a key process in the RAAS because angiotensin II causes the vasoconstriction association with hypertension. Because of its effectiveness as an ACE blocker, quinipril is widely used for clinical treatment of hypertension and chronic congestive heart failure(.) Matrix-assisted laser desorption/ionization coupled with time-of-flight analyzer (MALDI-TOF) is a high throughput instrument for biological sample analysis. This study developed a micro-scale approach for using MALDI-TOF to detect quinapril in biological samples. A micro-liquid-liquid-extraction strategy combined with ion-pair interaction successfully extracted quinapril from aqueous layer to organic layer. Quinolones were then used as matrix additives to suppress undesired substances in plasma produce signals. Several factors affecting extraction efficiency were investigated in a biosample with a volume of only 10 μL. This method is successful to monitor quinapril in the clinical therapeutic range. The proposed method proved effective for monitoring the trace amounts of quinapril typically used for clinical therapy. The relative standard deviation (R.S.D.) and relative error (R.E.) used for evaluating within- and between-day assays of quinapril in plasma consistently remained below 15%.
Product Description
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography
Supelco
Heptafluorobutyric acid, suitable for ion chromatography, ≥99.5% (GC)
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)