Merck
  • Examining histone posttranslational modification patterns by high-resolution mass spectrometry.
Examining histone posttranslational modification patterns by high-resolution mass spectrometry.
Methods in enzymology (2012-08-23)
Shu Lin, Benjamin A Garcia
ABSTRACT
Histone variants and posttranslational modifications (PTMs) are essential for epigenetic regulation of transcriptional expression. Single and/or combinatorial PTMs of histones play important roles in development and disease formation. Mass spectrometry (MS) has been a powerful tool to study histone variants and PTMs, as it not only can identify novel PTMs but also can provide quantitative measurement of a spectrum of histone variants and PTMs in the same sample. In this chapter, we employ a combination of chemical derivation and high-resolution MS to identify and quantify multiple histone variants and PTMs. Histones are acid extracted and modified with propionyl groups and subsequently produces suitable sizes of fragments for MS analysis by trypsin digestion. The newly generated N-termini of histone peptides can be differentially marked by stable isotope labeling in a second reaction of propionylation, which enables direct comparison between two different samples in the following MS analysis.
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N