Merck
  • Design, synthesis and pharmacological evaluation of omeprazole-like agents with anti-inflammatory activity.
Design, synthesis and pharmacological evaluation of omeprazole-like agents with anti-inflammatory activity.
Bioorganic & medicinal chemistry (2013-03-05)
Ahmed O H El-Nezhawy, Ayman R Biuomy, Fatma S Hassan, Ayman K Ismaiel, Hany A Omar
ABSTRACT
A new series of novel benzimidazole derivatives containing substituted pyrid-2-yl moiety and polyhydroxy sugar conjugated to the N-benzimidazole moiety has been synthesized and evaluated as orally bioavailable anti-inflammatory agents with anti-ulcerogenic activity. The anti-inflammatory and anti-ulcerogenic activities of these compounds were compared to diclofenac and omeprazole, respectively. In carrageenan-induced paw oedema assay, 2-methyl-N-((3,4-dimethoxypyridin-2-yl)methyl)-1H-benzimidazol-5-amine (12d) and 1-(1,2,3,5-tetrahydroxy-α-D-mannofuranose)-5-(((3,4-dimethoxypyridin-2yl)methyl)amino)-2-methyl-1H-benzimidazole (15d) displayed dose-dependent anti-inflammatory activities by decreasing the inflammation by 62% and 72%, respectively which is comparable to that of diclofenac (73%). In contrast to diclofenac, the anti-inflammatory activity of these compounds was not only free from any side effects on the gastric mucosa but also showed significant anti-ulcerogenic activity in rat pyloric ligation and ethanol-induced gastric ulcer models similar to that of omeprazole. Together, these findings suggest that 12d and 15d are potent anti-inflammatory agents with concurrent anti-ulcerogenic activity and support its clinical promise as a component of therapeutic strategies for inflammation, for which the gastric side effects are always a major limitation.
Sigma-Aldrich
Carrageenan, suitable for gel preparation
Sigma-Aldrich
2-Methylbenzimidazole, 98%
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Omeprazole, solid
Omeprazole, European Pharmacopoeia (EP) Reference Standard
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard