Merck
  • α-SNAP inhibits AMPK signaling to reduce mitochondrial biogenesis and dephosphorylates Thr172 in AMPKα in vitro.
α-SNAP inhibits AMPK signaling to reduce mitochondrial biogenesis and dephosphorylates Thr172 in AMPKα in vitro.
Nature communications (2013-03-07)
Lifu Wang, David L Brautigan
ABSTRACT
The AMP-activated protein kinase (AMPK) regulates metabolism in normal and pathological conditions and responds to nutrients, hormones, anti-diabetic drugs and physical exercise. AMPK is activated by the kinase LKB1 and inactivated by phosphatases whose identities remain uncertain. Here we show that AMPK associates with α-SNAP, an adapter that enables disassembly of cis-SNARE complexes formed during membrane fusion. Knockdown of α-SNAP activates AMPK to phosphorylate its endogenous substrates acetyl CoA carboxylase and Raptor, and provokes mitochondrial biogenesis. AMPK phosphorylation is rescued from α-SNAP RNA interference by LKB1 knockdown or expression of wild-type but not mutated α-SNAP. Recombinant wild-type but not mutated α-SNAP dephosphorylates pThr172 in AMPKα in vitro. Overexpression of wild-type but not mutated α-SNAP prevents AMPK activation in cells treated with agents to elevate AMP concentration. The mouse α-SNAP mutant hyh (hydrocephalus with hop gait) shows enhanced binding and inhibition of AMPK. By negatively controlling AMPK, α-SNAP therefore potentially coordinates membrane trafficking and metabolism.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material