Merck
  • dNTP-dependent conformational transitions in the fingers subdomain of Klentaq1 DNA polymerase: insights into the role of the "nucleotide-binding" state.
dNTP-dependent conformational transitions in the fingers subdomain of Klentaq1 DNA polymerase: insights into the role of the "nucleotide-binding" state.
The Journal of biological chemistry (2013-03-26)
Paul J Rothwell, William J Allen, Evangelos Sisamakis, Stanislav Kalinin, Suren Felekyan, Jerker Widengren, Gabriel Waksman, Claus A M Seidel
ABSTRACT
Conformational selection plays a key role in the polymerase cycle. Klentaq1 exists in conformational equilibrium between three states (open, closed, and “nucleotide-binding”) whose level of occupancy is determined by the bound substrate. The “nucleotide-binding” state plays a pivotal role in the reaction pathway. Direct evidence is provided for the role of a conformationally distinct “nucleotide-binding” state during dNTP incorporation. DNA polymerases are responsible for the accurate replication of DNA. Kinetic, single-molecule, and x-ray studies show that multiple conformational states are important for DNA polymerase fidelity. Using high precision FRET measurements, we show that Klentaq1 (the Klenow fragment of Thermus aquaticus DNA polymerase 1) is in equilibrium between three structurally distinct states. In the absence of nucleotide, the enzyme is mostly open, whereas in the presence of DNA and a correctly base-pairing dNTP, it re-equilibrates to a closed state. In the presence of a dNTP alone, with DNA and an incorrect dNTP, or in elevated MgCl2 concentrations, an intermediate state termed the "nucleotide-binding" state predominates. Photon distribution and hidden Markov modeling revealed fast dynamic and slow conformational processes occurring between all three states in a complex energy landscape suggesting a mechanism in which dNTP delivery is mediated by the nucleotide-binding state. After nucleotide binding, correct dNTPs are transported to the closed state, whereas incorrect dNTPs are delivered to the open state.
Sigma-Aldrich
2′-Deoxyadenosine 5′-triphosphate sodium salt solution, 10 mM
Pricing and availability is not currently available.
Sigma-Aldrich
2′-Deoxyadenosine 5′-triphosphate sodium salt solution, 100 mM, pH 7
Pricing and availability is not currently available.
Sigma-Aldrich
2′-Deoxyadenosine 5′-triphosphate disodium salt, ≥97%
Pricing and availability is not currently available.