Merck
  • [Estimating ability of dihydroquercetin to inhibit oxidation of fibrinogen by ozone].
[Estimating ability of dihydroquercetin to inhibit oxidation of fibrinogen by ozone].
Eksperimental'naia i klinicheskaia farmakologiia (2013-06-19)
V S Rogovskiĭ, M A Rozenfel'd, S D Razumovskiĭ, A I Matiushin, N L Shimanovskiĭ, B V Leonova, M L Konstantinova
ABSTRACT
The ability of dihydroquercetin to inhibit the oxidation of fibrinogen has been evaluated. It is established that dihydroquercetin inhibits oxidation of fibrinogen by ozone, thus preventing oxidative modification of fibrinogen and preserving its functional activity.
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Supelco
Taxifolin, analytical standard
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 35-65% protein (≥90% of protein is clottable).