Merck
  • Analysis of methylphosphonic acid, ethyl methylphosphonic acid and isopropyl methylphosphonic acid at low microgram per liter levels in groundwater.
Analysis of methylphosphonic acid, ethyl methylphosphonic acid and isopropyl methylphosphonic acid at low microgram per liter levels in groundwater.
Journal of chromatography. A (1998-01-22)
G A Sega, B A Tomkins, W H Griest
ABSTRACT
A method is described for determining methylphosphonic acid, ethyl methylphosphonic acid and isopropyl methylphosphonic acid, which are hydrolysis products of the nerve agents VX (S-2-diisopropylaminoethyl O-ethyl methylphosphonothiolate) and GB (sarin, isopropylmethyl phosphonofluoridate). The analytes are extracted from 50 ml groundwater using a solid-phase extraction column packed with 500 mg of silica with a bonded quaternary amine phase, and are eluted and derivatized with methanolic trimethylphenylammonium hydroxide. Separation and quantitation are achieved using a capillary column gas chromatograph equipped with a flame photometric detector operated in its phosphorus-selective mode. Two independent statistically-unbiased procedures were employed to determine the detection limits, which ranged between 3 and 9 micrograms/l, for the three analytes.
Sigma-Aldrich
Ethyl methylphosphonate, 98%