Merck
  • Pilot biomonitoring of adults and children following use of chlorpyrifos shampoo and flea collars on dogs.
Pilot biomonitoring of adults and children following use of chlorpyrifos shampoo and flea collars on dogs.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2011-01-18)
Melinda Bigelow Dyk, Zhenshan Chen, Sasan Mosadeghi, Helen Vega, Robert Krieger
ABSTRACT
Pesticide handlers and pet owners who use products such as shampoos and dips and insecticide-impregnated collars to treat and control fleas on companion animals are exposed to a variety of active ingredients. Chlorpyrifos exposures of adults and children were measured using urine biomonitoring following use of over-the-counter products on dogs. Age and gender-specific measurements of urinary 3, 5, 6-trichloro-2-pyridinol (TCPy) revealed modest elevations of biomarker excretion following shampoo/dips. Smaller TCPy increments were measured following application of impregnated dog collars. The extent of indoor activity and potential pet contact were important determinants of urine biomarker level. Children without direct pet contact excreted more TCPy following collar application. Pet collars may be a source of indoor surface contamination and human exposure. Children excreted up to 4 times more TCPy than adults when urine volumes were adjusted using age-specific creatinine excretion levels. Although chlorpyrifos is no longer used in the United States in pet care products, results of this research provide perspective on the extent of human exposure from similar pet care products. These pilot studies demonstrated that pet care products such as insecticidal shampoos and dips and impregnated collars may expose family members to low levels of insecticide relative to toxic levels of concern.
Product Description
Supelco
3,5,6-Trichloro-2-pyridinol, PESTANAL®, analytical standard