Merck
Metals and oxidative stress in patients with Parkinson's disease.
Annali dell'Istituto superiore di sanita (2005-10-26)
Giovanni Forte, Alessandro Alimonti, Anna Pino, Paolo Stanzione, Sonia Brescianini, Livia Brusa, Giuseppe Sancesario, Nicola Violante, Beatrice Bocca
ABSTRACT
Twenty-six metals and the oxidative status in 71 patients affected by Parkinson's disease and 44 healthy individuals were compared in order to identify potential biomarkers of the disease. In the patients, the following significant imbalances were found (p < or = 0.05): i) in serum, an increment of Ca, Mg, Ni, Si and V, and a decrement of Cd, Co, Fe, Li, Sn, Zn and Zr; ii) in blood, raised levels of Co, Li, Ni and Si and decreased of Al, Be, Ca, Cd, Fe, Mg, Mo, Sn, Zn and Zr; iii) increased formation of oxidant species and lowered anti-oxidant capacity (p < or = 0.001 for both). Barium, Bi, Cr, Cu, Hg, Mn, Pb, Sb, Sr, Tl and W did not change with the disease. The best discriminating variables between patients and controls were Cd, Co, Fe, Ni and Si in serum (91.2% of cases correctly classified), and Al, Cd, Co, Fe, Mo and Si in blood (98.2% of cases properly classified).
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Tungsten, foil, thickness 0.127 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis