Merck
  • Palladium-catalyzed annulation of arynes by o-halobenzamides: synthesis of phenanthridinones.
Palladium-catalyzed annulation of arynes by o-halobenzamides: synthesis of phenanthridinones.
The Journal of organic chemistry (2012-09-28)
Chun Lu, Anton V Dubrovskiy, Richard C Larock
ABSTRACT
The palladium-catalyzed annulation of arynes by substituted o-halobenzamides produces N-substituted phenanthridinones in good yields. This methodology provides this important heterocyclic ring system in a single step by simultaneous C-C and C-N bond formation, under relatively mild reaction conditions, and tolerates a variety of functional groups.
Sigma-Aldrich
2-Bromobenzamide, 98%
Pricing and availability is not currently available.