Merck
  • TGF-beta1-Induced MAPK activation promotes collagen synthesis, nodule formation, redox stress and cellular senescence in porcine aortic valve interstitial cells.
TGF-beta1-Induced MAPK activation promotes collagen synthesis, nodule formation, redox stress and cellular senescence in porcine aortic valve interstitial cells.
The Journal of heart valve disease (2014-01-05)
Dividutta Das, Andrew Holmes, Geraldine A Murphy, Kumaril Mishra, Anke C Rosenkranz, John D Horowitz, Jennifer A Kennedy
ABSTRACT
Aortic valve stenosis is a major cause of valve replacement, particularly in the elderly. TGF-beta1 is upregulated in stenotic valves and induces calcification and collagen synthesis in cultured valve interstitial cells. It has been shown previously that TGF-beta1 increases reactive oxygen species (ROS) in these cells in association with calcifying nodule formation, but the cellular signaling pathways responsible for these TGF-beta1-induced effects are not well defined. Cultured porcine aortic valve interstitial cells were used to investigate the effects of inhibitors of TGF-beta1 signaling pathways on 3H-proline incorporation into the extracellular matrix, the peak number of calcifying nodules formed, redox stress as dichlorofluorescein diacetate (DCF-DA) fluorescence, and senescence-associated beta-galactosidase staining. Nodule formation and proline incorporation were inhibited by SB431542, implicating the Smad pathway, by SB203580, implicating the P38 MAPK pathway, and by U0126, implicating the Mekl/2/Erk1/2 pathway in both processes. Fasudil, an inhibitor of the Rho kinase pathway, was selective in inhibiting nodule formation but not proline incorporation. It was verified that Smad2 phosphorylation, Erk1/2 phosphorylation and p38 MAPK phosphorylation were all induced by TGF-beta1, with Smad 2 phosphorylation peaking at 1-2 h and MAPK phosphorylation at 24-48 h. The effect of TGF-beta1 on phosphorylation of Smad 2 was inhibited by SB431542, on the phosphorylation of p38 MAPK was inhibited by SB203580, and on the phosphorylation of Erk1/2 was inhibited by U0126. ROS generation in response to TGF-beta1, measured as 2,7-dichlorofluorescein-diacetate fluorescence, was inhibited significantly by SB203580 and U0126, implicating both the p38 MAPK and Mekl/2/Erk1/2 signaling pathways. Both pathways also mediated TGF-beta1-induced cellular senescence which was localized to cellular aggregates and mature nodules. These data imply that the inhibition of either Smad or MAPK signaling pathways may have a therapeutic benefit in ameliorating the adverse pathological changes associated with aortic valve stenosis.
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder