Merck
  • Functionally diverse biotin-dependent enzymes with oxaloacetate decarboxylase activity.
Functionally diverse biotin-dependent enzymes with oxaloacetate decarboxylase activity.
Archives of biochemistry and biophysics (2013-11-05)
Adam D Lietzan, Martin St Maurice
ABSTRACT
Biotin-dependent enzymes catalyze carboxylation, decarboxylation and transcarboxylation reactions that participate in the primary metabolism of a wide range of organisms. In all cases, the overall reaction proceeds via two half reactions that take place in physically distinct active sites. In the first half-reaction, a carboxyl group is transferred to the 1-N' of a covalently tethered biotin cofactor. The tethered carboxybiotin intermediate subsequently translocates to a second active site where the carboxyl group is either transferred to an acceptor substrate or, in some bacteria and archaea, is decarboxylated to biotin and CO2 in order to power the export of sodium ions from the cytoplasm. A homologous carboxyltransferase domain is found in three enzymes that catalyze diverse overall reactions: carbon fixation by pyruvate carboxylase, decarboxylation and sodium transport by the biotin-dependent oxaloacetate decarboxylase complex, and transcarboxylation by transcarboxylase from Propionibacterium shermanii. Over the past several years, structural data have emerged which have greatly advanced the mechanistic description of these enzymes. This review assembles a uniform description of the carboxyltransferase domain structure and catalytic mechanism from recent studies of pyruvate carboxylase, oxaloacetate decarboxylase and transcarboxylase, three enzymes that utilize an analogous carboxyltransferase domain to catalyze the biotin-dependent decarboxylation of oxaloacetate.
Sigma-Aldrich
Pyruvate Carboxylase from bovine liver, buffered aqueous glycerol solution, 5-25 units/mg protein (BCA)
Sigma-Aldrich
Oxaloacetate Decarboxylase from Pseudomonas sp., lyophilized powder, ≥100 units/mg solid
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, meets USP testing specifications
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Biotin, United States Pharmacopeia (USP) Reference Standard
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SAFC
BIOTIN