Merck
  • Histatins: salivary peptides with copper(II)- and zinc(II)-binding motifs: perspectives for biomedical applications.
Histatins: salivary peptides with copper(II)- and zinc(II)-binding motifs: perspectives for biomedical applications.
The FEBS journal (2013-11-14)
Sonia Melino, Celeste Santone, Paolo Di Nardo, Bibudhendra Sarkar
ABSTRACT
Natural antimicrobial peptides represent a primordial mechanism of immunity in both vertebrate and nonvertebrate organisms. Among them, histatins belong to a family of human salivary metal-binding peptides displaying potent antibacterial, antifungal and wound-healing activities. These properties, along with the ability of histatins to inhibit collagenases and cysteine proteases, have attracted much attention for their potential use in the treatment of several oral diseases. This review critically assesses the studies carried out to date in order to provide a comprehensive and systematic vision of the information accumulated so far. In particular, the relationship between metal-binding and peptide activity is extensively analysed. The review provides important clues for developing possible therapeutic applications of histatins and their synthetic peptide analogues by creating a set of necessary resource materials to support investigators and industries interested in exploiting their unique properties.
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Copper, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.002mm, 2 micron polyesterimide insulation, 99.9%
Copper, tube, 100mm, outside diameter 1.75mm, inside diameter 0.33mm, wall thickness 0.71mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 12.7mm, inside diameter 10.26mm, wall thickness 1.22mm, as drawn, 99.9%
Copper, rod, 50mm, diameter 6.35mm, as drawn, 99.999%
Copper, tube, 1000mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Copper, tube, 100mm, outside diameter 12.7mm, inside diameter 10.26mm, wall thickness 1.22mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 0.95mm, inside diameter 0.29mm, wall thickness 0.33mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 1m coil, thickness 0.25mm, half hard, 99.95+%