Merck
  • The phylogeny of Ephemeroptera in Pterygota revealed by the mitochondrial genome of Siphluriscus chinensis (Hexapoda: Insecta).
The phylogeny of Ephemeroptera in Pterygota revealed by the mitochondrial genome of Siphluriscus chinensis (Hexapoda: Insecta).
Gene (2014-05-02)
Dan Li, Jia-Chang Qin, Chang-Fa Zhou
ABSTRACT
The mayfly species Siphluriscus chinensis (Siphluriscidae) has valuable structures useful for phylogeny reconstruction, given its putative basal position within the Ephemeroptera. Here its nearly complete mitochondrial genome is sequenced. We built phylogenetic trees through multiple analytical strategies with some other insect mitogenomes. Structurally, the obtained mitochondrial genome of S. chinensis is 16,616 bp in length,(1) containing 37 genes and an extra trnK-like (trnK2 (AAA)) gene. The 12 PCGs start with typical ATN codons, except the nad1 gene which starts with an unnormalized TTG. Like other known mayfly mitogenomes, the strand bias has negative AT-skew and negative GC-skew. Phylogenetically, our topologies suggest that Odonata is the basally diverged clade in Pterygota; Ephemeroptera is the sister group of the Neoptera; and S. chinensis is indeed the most basal mayfly branch.
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid