Merck
  • Complete mitochondrial genome of the atlas moth, Attacus atlas (Lepidoptera: Saturniidae) and the phylogenetic relationship of Saturniidae species.
Complete mitochondrial genome of the atlas moth, Attacus atlas (Lepidoptera: Saturniidae) and the phylogenetic relationship of Saturniidae species.
Gene (2014-05-07)
Miao-Miao Chen, Yan Li, Mo Chen, Huan Wang, Qun Li, Run-Xi Xia, Cai-Yun Zeng, Yu-Ping Li, Yan-Qun Liu, Li Qin
ABSTRACT
Mitochondrial genome (mitogenome) can provide information for genomic structure as well as for phylogenetic analysis and evolutionary biology. In this study, we present the complete mitogenome of the atlas moth, Attacus atlas (Lepidoptera: Saturniidae), a well-known silk-producing and ornamental insect with the largest wing surface area of all moths. The mitogenome of A. atlas is a circular molecule of 15,282 bp long, and its nucleotide composition shows heavily biased towards As and Ts, accounting for 79.30%. This genome comprises 13 protein-coding genes (PCGs), two ribosomal RNA genes (rRNAs), 22 transfer RNA genes (tRNAs), and an A+T-rich region. It is of note that this genome exhibits a slightly positive AT skew, which is different from the other known Saturniidae species. All PCGs are initiated by ATN codons, except for COI with CGA instead. Only six PCGs use a common stop codon of TAA or TAG, whereas the remaining seven use an incomplete termination codon T or TA. All tRNAs have the typical clover-leaf structure, with an exception for tRNA(Ser)(AGN). The A. atlas A+T-rich region contains non-repetitive sequences, but harbors several features common to the Bombycoidea insects. The phylogenetic relationships based on Maximum Likelihood method provide a well-supported outline of Saturniidae, which is in accordance with the traditional morphological classification and recent molecular works.
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder