Merck
  • Application of ANS fluorescent probes to identify hydrophobic sites on the surface of DREAM.
Application of ANS fluorescent probes to identify hydrophobic sites on the surface of DREAM.
Biochimica et biophysica acta (2014-05-24)
Walter G Gonzalez, Jaroslava Miksovska
ABSTRACT
DREAM (calsenilin or KChIP-3) is a calcium sensor involved in regulation of diverse physiological processes by interactions with multiple intracellular partners including DNA, Kv4 channels, and presenilin, however the detailed mechanism of the recognition of the intracellular partners remains unclear. To identify the surface hydrophobic surfaces on apo and Ca(2+)DREAM as a possible interaction sites for target proteins and/or specific regulators of DREAM function the binding interactions of 1,8-ANS and 2,6-ANS with DREAM were characterized by fluorescence and docking studies. Emission intensity of ANS-DREAM complexes increases upon Ca(2+) association which is consistent with an overall decrease in surface polarity. The dissociation constants for ANS binding to apoDREAM and Ca(2+)DREAM were determined to be 195±20μM and 62±4μM, respectively. Fluorescence lifetime measurements indicate that two ANS molecules bind in two independent binding sites on DREAM monomer. One site is near the exiting helix of EF-4 and the second site is located in the hydrophobic crevice between EF-3 and EF-4. 1,8-ANS displacement studies using arachidonic acid demonstrate that the hydrophobic crevice between EF-3 and EF-4 serves as a binding site for fatty acids that modulate functional properties of Kv4 channel:KChIP complexes. Thus, the C-terminal hydrophobic crevice may be involved in DREAM interactions with small hydrophobic ligands as well as other intracellular proteins.
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, suitable for fluorescence, ≥95.0% (T)
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, suitable for fluorescence, ≥95% (TLC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, technical, ≥90% (NT)