Merck
  • Intrathecal application of the nimodipine slow-release microparticle system eg-1962 for prevention of delayed cerebral ischemia and improvement of outcome after aneurysmal subarachnoid hemorrhage.
Intrathecal application of the nimodipine slow-release microparticle system eg-1962 for prevention of delayed cerebral ischemia and improvement of outcome after aneurysmal subarachnoid hemorrhage.
Acta neurochirurgica. Supplement (2014-11-05)
Nima Etminan, R Loch Macdonald, Cara Davis, Kevin Burton, Hans-Jakob Steiger, Daniel Hänggi
ABSTRACT
The effective reduction of delayed cerebral ischemia (DCI), a main contributor for poor outcome following aneurysmal subarachnoid hemorrhage (SAH), remains challenging. Previous clinical trials on systemic pharmaceutical treatment of SAH mostly failed to improve outcome, probably because of insensitive pharmaceutical targets and outcome measures, small sample size, insufficient subarachnoid drug concentrations and also detrimental, systemic effects of the experimental treatment per se. Interestingly, in studies that are more recent, intrathecal administration of nicardipine pellets following surgical aneurysm repair was suggested to have a beneficial effect on DCI and neurological outcome. However, this positive effect remained restricted to patients who were treated surgically for a ruptured aneurysm. Because of the favorable results of the preclinical data on DCI and neurological outcome in the absence of neurotoxicity or systemic side effects, we are initiating clinical trials. The PROMISE (Prolonged Release nimOdipine MIcro particles after Subarachnoid hemorrhage) trial is designed as an unblinded, nonrandomized, single-center, single-dose, dose-escalation safety and tolerability phase 1 study in patients surgically treated for aSAH and will investigate the effect of intracisternal EG-1962 administration. The NEWTON (Nimodipine microparticles to Enhance recovery While reducing TOxicity after subarachNoid hemorrhage) trial is a phase 1/2a multicenter, controlled, randomized, open-label, dose-escalation, safety, tolerability, and pharmacokinetic study comparing EG-1962 and nimodipine in patients with aneurysmal SAH.
Product Description
Supelco
Nimodipine, Pharmaceutical Secondary Standard; Certified Reference Material
Nimodipine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nimodipine