Merck
  • Role of dermal exposure in systemic intake of methylenediphenyl diisocyanate (MDI) among construction and boat building workers.
Role of dermal exposure in systemic intake of methylenediphenyl diisocyanate (MDI) among construction and boat building workers.
Toxicology letters (2014-12-30)
Maj-Len Henriks-Eckerman, Erja A Mäkelä, Juha Laitinen, Katriina Ylinen, Katri Suuronen, Aki Vuokko, Riitta Sauni
ABSTRACT
The causal relationship between inhalation exposure to methylenediphenyl diisocyanate (MDI) and the risk of occupational asthma is well known, but the role of dermal exposure and dermal uptake of MDI in this process is still unclear. The aims of this study were to measure dermal exposure to and the dermal uptake of MDI among workers (n=24) who regularly handle MDI-urethanes. Dermal exposure was measured by the tape-strip technique from four sites on the dominant hand and arm. The workers with the highest exposure (n=5) were biomonitored immediately after their work shift, in the evening and the next morning, using urinary 4,4´methylenedianiline (MDA) as a marker. Dermal uptake was evaluated by comparing workers' MDA excretions both when they were equipped with respiratory protective devices (RPDs) and when they did not use them. The measured amounts of MDI on their hands varied from below 0.1 to 17 μg/10 cm(2) during the test. MDI concentrations were in the range of 0.08 to 27 μg m(-3) in the breathing zone outside the RPDs. MDA concentrations varied from 0.1 to 0.2 μmol mol(-1) creatinine during the test period. The decreasing effect of RPDs on inhalation exposure was absent in the next morning urine samples; this excretion pattern might be an indication of dermal uptake of MDI.
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Supelco
Acetic anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Heptafluorobutyric anhydride, ≥98%