Merck
  • Effect of volatile oil from Blumea Balsamifera (L.) DC. leaves on wound healing in mice.
Effect of volatile oil from Blumea Balsamifera (L.) DC. leaves on wound healing in mice.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan (2015-01-27)
Yuxin Pang, Dan Wang, Xuan Hu, Hui Wang, Wanjin Fu, Zuowang Fan, Xiaolu Chen, Fulai Yu
ABSTRACT
To assess the effectiveness of violate oil from Blumea Balsamifera (L.) DC. leaves (BB oil) on wound healing in mice. Undiluted BB oil and its diluted solutions with olive oil to 1/5 and 1/10 to yield BB oil1/5 and BB oil1/10 were applied to the wounded skin before wound healing conditions were assessed by healing rate, histopathology, and contents of collagen, hydroxyproline, and Neuropeptide Substance P (SP). All above results were compared with the efficacies of the control, pure olive oil, basic fibroblast growth factor (BFGF), and cream of Jing Wan Hong (JWH). BB oil1/5 and BB oil1/10 improved wound contraction and closure. Histopathology study further confirmed a desirable histological organization of wound tissues. BB oil1/5 and BB oil1/10 reduced the number of inflammatory cells, increased wound-healing rates, and significantly increased the hydroxyproline content. Both BB oil1/5 and BB oil1/10 improved formation of collagen, and reduced the frequency of fibroblasts. Moreover, BB oil1/5 and BB oil1/10 markedly promoted SP expression. However, undiluted BB oil may induce skin thickening and hardening, inhibite collagen synthesis and delay complete skin wound healing. The BB oil1/5 and BB oil1/10 promoted capillary regeneration, blood circulation, collagen deposition, granular tissue formation, epithelial deposition, and wound contraction. The mechanism underlying the action might be related to induction of SP secretion, and the proliferation and differentiation of mesenchymal cells.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered