Merck
  • Free fatty acid levels in fluid of dominant follicles at the preferred insemination time in dairy cows are not affected by early postpartum fatty acid stress.
Free fatty acid levels in fluid of dominant follicles at the preferred insemination time in dairy cows are not affected by early postpartum fatty acid stress.
Journal of dairy science (2015-02-05)
Hilde Aardema, Bart M Gadella, Chris H A van de Lest, Jos F H M Brouwers, Tom A E Stout, Bernard A J Roelen, Peter L A M Vos
ABSTRACT
The fertility of high-yielding dairy cows has declined during the last 3 decades, in association with a more profound negative energy balance (NEB) during the early weeks postpartum. One feature of this NEB is a marked elevation in circulating free fatty acid (FFA) concentrations. During the early postpartum period (≤ d 42), circulatory FFA levels were measured weekly, and progesterone concentrations and the diameter of the dominant follicles were determined thrice weekly. Retrospectively, cows that ovulated within 35 d postpartum were grouped as "normal ovulating" cows (n = 5), and the others were grouped as "delayed ovulating" cows (n = 5). In both groups, high total FFA levels (>500 µM) were evident immediately postpartum. Interestingly, cows with delayed ovulation had higher plasma FFA concentrations in the first weeks postpartum compared with normal ovulating cows. In both cow groups, FFA decreased to control levels of non-NEB cows within 3 wk postpartum. The FFA compositions and concentrations in fluids from the dominant follicles of postpartum cows were not different between the normal and delayed ovulating cows when measured at potential insemination points: d 55, 80, and 105 postpartum. Interestingly, the concentration of monounsaturated oleic acid was higher and that of saturated stearic acid lower in follicular fluids of both groups compared with that in blood. The level of FFA in follicular fluid was correlated with the ratio of 17β-estradiol (E2) to progesterone (P4) in follicular fluid, with a relatively high level of unsaturated FFA in follicles with a low E2:P4 ratio. Taken together, these results indicate that a more severe NEB early postpartum is related to a delay in the first postpartum ovulation and does not affect FFA composition in follicular fluid at the preferred insemination time. The high FFA level in dominant follicles with a low E2:P4 ratio may be due to a different FFA metabolism in these follicles. The diagnostic value of this observation for selective screening of dominant follicles needs further investigation.
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Palmitic acid, ≥99%
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Palmitic acid, natural, 98%, FG