Merck
  • Characterization of a Kunitz-type protease inhibitor peptide (Rusvikunin) purified from Daboia russelii russelii venom.
Characterization of a Kunitz-type protease inhibitor peptide (Rusvikunin) purified from Daboia russelii russelii venom.
International journal of biological macromolecules (2014-03-19)
Ashis K Mukherjee, Stephen P Mackessy, Sumita Dutta
ABSTRACT
The snake venom may be considered as a potent source of untapped therapeutic proteins and peptides. The peptide mass fingerprinting and N-terminal sequence alignment of a 6.9kDa peptide named Rusvikunin from Daboia russelii russelii venom show the presence of putative conserved domains of the KU superfamily. Further, BLAST analysis of two of the de novo peptide sequences of Rusvikunin demonstrates significant sequence homology with serine proteases reported in the NCBI database. Rusvikunin possesses conserved cysteine residues and Arg15 at the P1 position. It inhibits amidolytic activity of trypsin (IC50=50nmol/l), plasmin (IC50=1.1μmol/l), and fibrinogen clotting as well as plasma clotting activity of thrombin (IC50=1.3μmol/l); however, it does not inhibit the amidolytic activity of chymotrypsin, thrombin, factor Xa, and tissue plasminogen activator. Rusvikunin is a glycoprotein, demonstrates dose-dependent BAEE-esterase activity. It does not show lethality in mice or in vitro cytotoxicity against mammalian cells but shows in vivo anticoagulant activity 6h after i.p. injection in the mouse model. The commercial polyvalent and monovalent antivenom failed to inhibit the functional properties of Rusvikunin. The possible biomedical applications of Rusvikunin in the treatment and/or prevention of cardiovascular disorders such as thrombosis and trypsin-induced inflammation are suggested.
Product Description
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS