Merck
  • MeCP2 deficiency is associated with reduced levels of tubulin acetylation and can be restored using HDAC6 inhibitors.
MeCP2 deficiency is associated with reduced levels of tubulin acetylation and can be restored using HDAC6 inhibitors.
Journal of molecular medicine (Berlin, Germany) (2014-09-12)
W A Gold, T A Lacina, L C Cantrill, John Christodoulou
ABSTRACT
Rett syndrome (RTT) is a severe neurodevelopmental disorder, predominantly caused by loss of function mutations in the X-linked methyl-CpG-binding protein 2 (MECP2) gene. Despite the genetic cause being known in the majority of cases, the pathophysiology of the neurological phenotype of RTT is largely unknown. Tubulin and the microtubule network play an essential role in neuronal function whereby the acetylation state of microtubules dictates the efficiency of neuronal migration and differentiation, synaptic targeting and molecular motor trafficking of mRNA, high-energy mitochondria and brain-derived neurotrophic factor (BDNF)-containing vesicles. Recent reports have shown perturbations in tubulin and microtubule dynamics in MeCP2-deficient cells, suggesting a link between the aberrations of these cellular entities and the neurobiology of RTT. We have interrogated the functional state of the microtubule network in fibroblasts derived from two patients with RTT as well as cortical neurons from a RTT mouse model and observed a reduction in acetylated α-tubulin and an increase in the tubulin-specific deacetylase, histone deacetylase 6 (HDAC6). Furthermore, we show that inhibition of HDAC6 by Tubastatin A can restore tubulin acetylation levels. We also demonstrate microtubule instability in the RTT patient fibroblasts in response to nocodazole, which is progressively ameliorated in a mutation-dependent manner by Tubastatin A. We conclude that Tubastatin A is capable of counteracting the microtubule defects observed in MeCP2-deficient cells, which could in turn lead to the restoration of molecular trafficking along the microtubules and thus could be a potentially new therapeutic option for RTT. Cells from MeCP2-deficient cells show reduced levels of acetylated α-tubulin. Cells from two patients and a RTT mouse model have increased levels of HDAC6 but not sirtuin 2 (SIRT2). Inhibition of HDAC6 by Tubastatin A increases the in vitro acetylation of α-tubulin. Inhibition of HDAC6 by Tubastatin A does not increase MECP2 expression. Cells from two patients show microtubule instability, which is ameliorated by Tubastatin A.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture