Merck
  • Quantification of recombinant human erythropoietin by amino acid analysis using isotope dilution liquid chromatography-tandem mass spectrometry.
Quantification of recombinant human erythropoietin by amino acid analysis using isotope dilution liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2014-05-21)
Jung-Hyuk Yim, Ina Yoon, Hyo-Jin Yang, Sook-Kyung Kim, Sang-Ryoul Park, Yong-Moon Lee, Ji-Seon Jeong
ABSTRACT
Herein, we describe an accurate method for protein quantification based on conventional acid hydrolysis and an isotope dilution-ultra performance liquid chromatography-tandem mass spectrometry method. The analyte protein, recombinant human erythropoietin (rhEPO), was effectively hydrolyzed by incubation with 8 mol/L hydrochloric acid at 130 °C for 48 h, in which at least 1 μmol/kg of rhEPO was treated to avoid possible degradation of released amino acids during hydrolysis. Prior to hydrolysis, sample solution was subjected to ultrafiltration to eliminate potential interfering substances. In a reversed-phase column, the analytes (phenylalanine, proline, and valine) were separated within 3 min using gradient elution comprising 20 % (v/v) acetonitrile and 10 mmol/L ammonium acetate, both containing 0.3 % (v/v) trifluoroacetic acid. The optimized hydrolysis and analytical conditions in our study were strictly validated in terms of accuracy and precision, and were suitable for the accurate quantification of rhEPO. Certified rhEPO was analyzed using a conventional biochemical assay kit as an additional working calibrant for the quantification of EPO and improved the accuracy. The optimized protocol is suitable for the accurate quantification of rhEPO and satisfactorily serves as a reference analytical procedure for the certification of rhEPO and similar proteins.
Product Description
SAFC
L-Isoleucine
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Supelco
Acetonitrile, analytical standard
Supelco
Trifluoroacetic acid, analytical standard
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%