Merck
  • 8-Benzyltetrahydropyrazino[2,1-f]purinediones: water-soluble tricyclic xanthine derivatives as multitarget drugs for neurodegenerative diseases.
8-Benzyltetrahydropyrazino[2,1-f]purinediones: water-soluble tricyclic xanthine derivatives as multitarget drugs for neurodegenerative diseases.
ChemMedChem (2014-05-13)
Andreas Brunschweiger, Pierre Koch, Miriam Schlenk, Felipe Pineda, Petra Küppers, Sonja Hinz, Meryem Köse, Stefan Ullrich, Jörg Hockemeyer, Michael Wiese, Jag Heer, Christa E Müller
ABSTRACT
8-Benzyl-substituted tetrahydropyrazino[2,1-f]purinediones were designed as tricyclic xanthine derivatives containing a basic nitrogen atom in the tetrahydropyrazine ring to improve water solubility. A library of 69 derivatives was prepared and evaluated in radioligand binding studies at adenosine receptor (AR) subtypes and for their ability to inhibit monoamine oxidases (MAO). Potent dual-target-directed A1 /A2A adenosine receptor antagonists were identified. Several compounds showed triple-target inhibition; one of the best compounds was 8-(2,4-dichloro-5-fluorobenzyl)-1,3-dimethyl-6,7,8,9-tetrahydropyrazino[2,1-f]purine-2,4(1H,3H)-dione (72) (human AR: Ki  A1 217 nM, A2A 233 nM; IC50 MAO-B: 508 nM). Dichlorinated compound 36 [8-(3,4-dichlorobenzyl)-1,3-dimethyl-6,7,8,9-tetrahydropyrazino[2,1-f]purine-2,4(1H,3H)-dione] was found to be the best triple-target drug in rat (Ki  A1 351 nM, A2A 322 nm; IC50 MAO-B: 260 nM), and may serve as a useful tool for preclinical proof-of-principle studies. Compounds that act at multiple targets relevant for symptomatic as well as disease-modifying treatment of neurodegenerative diseases are expected to show advantages over single-target therapeutics.
Sigma-Aldrich
Monoamine Oxidase B human, recombinant, expressed in baculovirus infected BTI insect cells
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Monoamine Oxidase A human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Adenosine