Merck
  • UHPLC-UV method for the determination of flavonoids in dietary supplements and for evaluation of their antioxidant activities.
UHPLC-UV method for the determination of flavonoids in dietary supplements and for evaluation of their antioxidant activities.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Sylwia Magiera, Irena Baranowska, Anna Lautenszleger
ABSTRACT
A simple and rapid ultra-high performance liquid chromatographic (UHPLC) method coupled with an ultraviolet detector (UV) has been developed and validated for the separation and determination of 14 major flavonoids ((±)-catechin, (-)-epicatechin, glycitin, (-)-epicatechin gallate, rutin, quercitrin, hesperidine, neohesperidine, daidzein, glycitein, quercetin, genistein, hesperetin, and biochanin A) in herbal dietary supplements. The flavonoids have been separated on a Chromolith Fast Gradient Monolithic RP-18e column utilizing a mobile phase composed of 0.05% trifluoroacetic acid in water and acetonitrile in gradient elution mode. Under these conditions, flavonoids were separated in a 5 min run. The selectivity of the developed UHPLC-UV method was confirmed by comparison with ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) analysis. The validation parameters such as linearity, sensitivity, precision, and accuracy were found to be highly satisfactory. The optimized method was applied to determination of flavonoids in different dietary supplements. Additionally, the developed HPLC-UV method combined with 2,2-diphenyl-1-picrylhydrazyl radical (DPPH) assay was used in the evaluation of antioxidant activity of the selected flavonoids.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Chrysin, ≥96.5%
Supelco
Chrysin, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Neohesperidin, ≥90% (HPLC)
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Supelco
Hesperidin, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Daidzein, analytical standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Hesperidin, primary reference standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard