Merck
  • Inactivation of HDAC1 or HDAC2 induces gamma globin expression without altering cell cycle or proliferation.
Inactivation of HDAC1 or HDAC2 induces gamma globin expression without altering cell cycle or proliferation.
American journal of hematology (2015-03-27)
Erica B Esrick, Marie McConkey, Katherine Lin, Alyse Frisbee, Benjamin L Ebert
ABSTRACT
Other than hydroxyurea, no pharmacologic agents are clinically available for fetal hemoglobin (HbF) induction in sickle cell disease (SCD). An optimal candidate would induce HbF without causing cell cycle inhibition and would act independently of hydroxyurea in order to yield additional HbF induction when combined. We explored whether inhibition of histone deacetylase (HDAC) 1 or HDAC2 could achieve these goals. In human erythroid progenitor cells, shRNA knockdown of the HDAC1 or HDAC2 genes induced gamma globin, without altering cellular proliferation in vitro, and without altering cell cycle phase. Treatment with hydroxyurea in combination with HDAC2 knockdown yielded a further increase in gamma globin expression. Additionally, when CD34+ cells were treated with both hydroxyurea and MS-275 (an inhibitor of HDAC 1, 2, and 3), an additive induction of relative gamma globin expression was achieved. Our findings support further clinical investigation of HDAC inhibitors in combination with hydroxyurea in SCD patients.
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder