Merck
  • Role of polyol moiety of amphotericin B in ion channel formation and sterol selectivity in bilayer membrane.
Role of polyol moiety of amphotericin B in ion channel formation and sterol selectivity in bilayer membrane.
Bioorganic & medicinal chemistry (2015-07-26)
Tomoya Yamamoto, Yuichi Umegawa, Hiroshi Tsuchikawa, Nobuaki Matsumori, Shinya Hanashima, Michio Murata, Resul Haser, Bernard J Rawlings, Patrick Caffrey
ABSTRACT
Amphotericin B (AmB) is a polyene macrolide antibiotic widely used to treat mycotic infections. In this paper, we focus on the role of the polyol moiety of AmB in sterol selectivity using 7-oxo-AmB, 7α-OH-AmB, and 7β-OH-AmB. The 7-OH analogs were prepared from 7-oxo-AmB. Their K(+) flux activity in liposomes showed that introduction of an additional ketone or hydroxy group on the polyol moiety reduces the original activity. Conformational analyses of these derivatives indicated that intramolecular hydrogen-bonding network possibly influenced the conformational rigidity of the macrolactone ring, and stabilized the active conformation in the membrane. Additionally, the flexible polyol leads to destabilization of the whole macrolactone ring conformation, resulting in a loss of sterol selectivity.
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Valinomycin, ≥99.0% (TLC)
Sigma-Aldrich
2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein, ≥90% (HPLC)
Sigma-Aldrich
Ergosterol, ≥75%
Sigma-Aldrich
Valinomycin, ≥98% (TLC), ≥90% (HPLC)
Sigma-Aldrich
Valinomycin, Ready Made Solution, ~1 mg/mL in DMSO
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥95.5% (GC), ≥98% (TLC)
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥99.0% (TLC)
SAFC
MOPS
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications