Merck
  • An optimized method for high-titer lentivirus preparations without ultracentrifugation.
An optimized method for high-titer lentivirus preparations without ultracentrifugation.
Scientific reports (2015-09-09)
Wei Jiang, Rui Hua, Mengping Wei, Chenhong Li, Zilong Qiu, Xiaofei Yang, Chen Zhang
ABSTRACT
Lentiviral technology has proven to be a powerful tool to express exogenous genes in dividing and non-dividing cells. Currently, most protocols for generating high-titer lentivirus require ultracentrifugation, which can be an instrumental barrier for routine operations in a laboratory. In this study, the effect of relative centrifugal force (RCF) on the concentration efficiency of the lentivirus was systematically explored, and it was found that sucrose gradient centrifugation with a relatively low speed (≤10,000 g) robustly produces a high-titer virus (up to 2×10(8) TU/ml). The optimal sucrose concentration is 10%, and the recovery rate of the functional virus is greater than 80%. The infection efficiency of both concentrated and un-concentrated lentivirus decreases rapidly when the viruses are stored at 4 °C (τ≈1.3 days) or subjected to multiple freeze-thaw cycles (τ=1.1 rounds). In summary, we describe an efficient and easy-to-handle protocol for high-titer lentivirus purification.
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis