Merck
  • Modulation of Electron Injection Dynamics of Ru-Based Dye/TiO2 System in the Presence of Three Different Organic Solvents: Role of Solvent Dipole Moment and Donor Number.
Modulation of Electron Injection Dynamics of Ru-Based Dye/TiO2 System in the Presence of Three Different Organic Solvents: Role of Solvent Dipole Moment and Donor Number.
Chemphyschem : a European journal of chemical physics and physical chemistry (2015-04-03)
Subrata Mahanta, Hiroyuki Matsuzaki, Takurou N Murakami, Ryuzi Katoh, Hajime Matsumoto, Akihiro Furube
ABSTRACT
In the present work, femtosecond transient absorption spectroscopy (fs-TAS) has been employed to investigate the electron injection efficiency (EIE) both from the singlet and triplet excited states of a well-known ruthenium dye (N719) to the conduction band (CB) of nanostructured TiO(2) in presence of three different organic solvents [γ-butylactone (GBL), 3-methoxypropionitrile (MPN), and dimethylformamide (DMF)] with different donor numbers (DNs) and dipole moments (DMs). The DM and DN of a solvent modulates the CB edge energy of TiO(2), and this effect reflects well in the fs-TAS results, which shows an EIE trend following the order GBL≥MPN≫DMF, that is, highest in GBL and lowest in DMF solvent environments. Fs-TAS results indicate a lower contribution of electron injection from both the singlet and triplet states in DMF, for which the dominant adsorption of DMF molecules on the TiO(2) surface seems to play an important role in the mechanism.
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%