Merck
  • Characterization and identification of the xylanolytic enzymes from Aspergillus fumigatus Z5.
Characterization and identification of the xylanolytic enzymes from Aspergillus fumigatus Z5.
BMC microbiology (2015-06-24)
Youzhi Miao, Juan Li, Zhizhuang Xiao, Qirong Shen, Ruifu Zhang
ABSTRACT
Plant biomass, the most abundant natural material on earth, represents a vast source of food and energy in nature. As the main component of plant biomass, xylan is a complex polysaccharide comprising a linear β(1,4)-linked backbone of xylosyl residues substituted by acetyl, arabinosyl, glucuronysyl and 4-O-methylglucuronycyl residues. Aspergillus fumigatus Z5 is an efficient plant biomass depolymerization fungus. In this study, its crude xylanolytic enzymes were characterized and identified by two-dimensional gel electrophoresis (2-DE). The optimal temperature for the crude xylanases was close to 60 °C, the highest xylanase activity was achieved at pH ranged from 3 to 6, and the crude xylanases also showed a very broad region of pH (3-11) stability. The maximal xylanase activity of 21.45 U · ml(-1) was observed in the fourth day of cultivation at 50 °C and 150 rpm with 2 % xylan as the sole carbon source. Zymogram analysis indicated that there were more than seven secreted proteins with xylanase activity. In the crude enzyme, two major endoxylanases, five cellulases and several associated enzymes were identified to be involved in the hydrolysis of polysaccharides. Of the total 13 xylanase genes in the Z5 genome, 11 were observed using q-PCR to be induced by xylan, one of which, An endo-1,4-β-xylanase with a low secretion level, was also expressed and characterized. The final hydrolysis products of xylan by crude enzyme mainly consisted of xylobiose. This study provides a comprehensive understanding of the depolymerization of xylan by Z5 and will help to design enzymatic strategies for plant biomass utilization.
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra