Merck
Presumed pseudokinase VRK3 functions as a BAF kinase.
Biochimica et biophysica acta (2015-04-23)
Choon-Ho Park, Hye Guk Ryu, Seong-Hoon Kim, Dohyun Lee, Haengjin Song, Kyong-Tai Kim
ABSTRACT
Vaccinia-related kinase 3 (VRK3) is known as a pseudokinase that is catalytically inactive due to changes in motifs that are essential for kinase activity. Although VRK3 has been regarded as a genuine pseudokinase from structural and biochemical studies, recent reports suggest that VRK3 acts as an active kinase as well as a signaling scaffold in cells. Here, we demonstrate that VRK3 phosphorylates the nuclear envelope protein barrier-to-autointegration factor (BAF) on Ser4. Interestingly, VRK3 kinase activity is dependent upon its N-terminal regulatory region, which is excluded from the determination of its crystal structure. Furthermore, the kinase activity of VRK3 is involved in the regulation of the cell cycle. VRK3 expression levels increase during interphase, whereas VRK1 is enriched in late G2 and early M phase. Ectopic expression of VRK3 induces the translocation of BAF from the nucleus to the cytoplasm. In addition, depletion of VRK3 decreases the population of proliferating cells. These data suggest that VRK3-mediated phosphorylation of BAF may facilitate DNA replication or gene expression by facilitating the dissociation of nuclear envelope proteins and chromatin during interphase.
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%