Merck
  • The expression of CDK1 is associated with proliferation and can be a prognostic factor in epithelial ovarian cancer.
The expression of CDK1 is associated with proliferation and can be a prognostic factor in epithelial ovarian cancer.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-04-26)
Qinghua Xi, Menghui Huang, Yingying Wang, Jianxin Zhong, Rong Liu, Guiqin Xu, Lifei Jiang, Juan Wang, Zheng Fang, Shuyun Yang
ABSTRACT
Overexpression of cyclin-dependent kinase 1 (CDK1) has been noted to correlation with several human cancers. However, the effects of CDK1 on ovarian cancer development remain unclear. The aim of this study was to examine the effect of CDK1 and related mechanism in the proliferation and resistance to chemotherapeutic drugs of epithelial ovarian cancer (EOC). Immunohistochemical analysis was performed in 119 human ovarian cancer samples, and the data were correlated with clinicopathologic features. Furthermore, Western blot analysis was performed for CDK1 in EOC samples and cell lines to evaluate their protein levels and molecular interaction. Kaplan-Meier survival analysis showed that strong expression of CDK1 exhibited a significant correlation with poor prognosis in human EOC (P = 0.02). Meanwhile, we found that knockdown CDK1 by shCDK1 promoted the apoptosis rate and increased the sensitivity to chemotherapy drugs. Thus, CDK1 might serve as a prognostic marker, and it might be of great value for experimental therapies in EOC.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cdc2a
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Propidium iodide solution