Merck
  • Predicting Dissolved Inorganic Carbon in Photoautotrophic Microalgae Culture via the Nitrogen Source.
Predicting Dissolved Inorganic Carbon in Photoautotrophic Microalgae Culture via the Nitrogen Source.
Environmental science & technology (2015-07-29)
Binh T Nguyen, Bruce E Rittmann
ABSTRACT
Dissolved inorganic carbon (DIC) and pH are key factors that control the growth rate of microalgae growing photoautotrophically. Being able to quantify how DIC and pH independently affect growth kinetics requires a means to control each parameter independently. In this study, we used the Proton Condition (PC) to develop means to control pH and DIC independently. Using the PC, we found that different N sources systematically affect the alkalinity and the DIC in distinct ways. With pH controlled at a fixed level by CO2 addition, using nitrate as the N source increased the alkalinity and DIC concentration in proportion to the increase in biomass concentration. In contrast, using ammonium caused the alkalinity and DIC to decline, while using ammonium nitrate left the DIC nearly unchanged. Experiments with a model photoautotroph cyanobacterium, Synechocystis sp. PCC 6803, in batch experiments with modified BG-11 media and a pH-stat confirmed all of the DIC predictions of the PC-based model. Thus, this study provides a mechanistic basis for managing the DIC for photoautotrophic cultures through the N source. In particular, using ammonium nitrate makes it possible to control DIC and pH independently in a pH-stat.
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C