Merck
Aspergillus species as emerging causative agents of onychomycosis.
Journal de mycologie medicale (2015-02-07)
S Nouripour-Sisakht, H Mirhendi, M R Shidfar, B Ahmadi, A Rezaei-Matehkolaei, M Geramishoar, F Zarei, N Jalalizand
ABSTRACT
Onychomycosis is a common nail infection caused by dermatophytes, non-dermatophyte molds (NDM), and yeasts. Aspergillus species are emerging as increasing causes of toenail onychomycosis. The purpose of this study was species delineation of Aspergillus spp. isolated from patients with onychomycosis. During a period of one year (2012-2013), nail samples were collected from patients clinically suspected of onychomycosis and subjected to microscopic examination and culture. Species identification was performed based on macro- and micro-morphology of colonies. For precise species identification, PCR-amplification and sequencing of the beta-tubulin gene followed by BLAST queries were performed where required. A total of 463/2,292 (20.2%) tested nails were diagnosed with onychomycosis. Among the positive specimens, 154 cases (33.2%) were identified as saprophytic NDM onychomycosis, 135 (29.2%) of which were attributable to Aspergillus. Aspergillus species isolated from the infected nails included Aspergillus flavus (77.3%, n=119), Aspergillus niger (n=4), Aspergillus tubingensis (n=4), Aspergillus terreus (n=3), Aspergillus sydowii (n=2), Aspergillus spp. (n=2), and Aspergillus candidus (n=1). Among the patients diagnosed with onychomycosis due to Aspergillus (average patient age, 47.4 years), 40 had fingernail and 95 toenail involvement. The large toenails were most commonly affected. This study identified a markedly high occurrence of A. flavus, and this fungus appears to be an emerging cause of saprophytic onychomycosis in Iran. The study moreover highlights the necessity of differentiating between dermatophytic and non-dermatophytic nail infections for informed decisions on appropriate therapy.
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, 98%
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Boric acid, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, BioUltra, Molecular Biology, ≥99.5% (T)