Merck
  • Vaccinia extracellular virions enter cells by macropinocytosis and acid-activated membrane rupture.
Vaccinia extracellular virions enter cells by macropinocytosis and acid-activated membrane rupture.
The EMBO journal (2011-07-28)
Florian Ingo Schmidt, Christopher Karl Ernst Bleck, Ari Helenius, Jason Mercer
ABSTRACT
Vaccinia virus (VACV), the model poxvirus, produces two types of infectious particles: mature virions (MVs) and extracellular virions (EVs). EV particles possess two membranes and therefore require an unusual cellular entry mechanism. By a combination of fluorescence and electron microscopy as well as flow cytometry, we investigated the cellular processes that EVs required to infect HeLa cells. We found that EV particles were endocytosed, and that internalization and infection depended on actin rearrangements, activity of Na(+)/H(+) exchangers, and signalling events typical for the macropinocytic mechanism of endocytosis. To promote their internalization, EVs were capable of actively triggering macropinocytosis. EV infection also required vacuolar acidification, and acid exposure in endocytic vacuoles was needed to disrupt the outer EV membrane. Once exposed, the underlying MV-like particle presumably fused its single membrane with the limiting vacuolar membrane. Release of the viral core into the host cell cytosol allowed for productive infection.
Sigma-Aldrich
IPA-3, ≥95% (HPLC), powder
Pricing and availability is not currently available.