Merck
  • Significantly enhanced tumor cellular and lysosomal hydroxychloroquine delivery by smart liposomes for optimal autophagy inhibition and improved antitumor efficiency with liposomal doxorubicin.
Significantly enhanced tumor cellular and lysosomal hydroxychloroquine delivery by smart liposomes for optimal autophagy inhibition and improved antitumor efficiency with liposomal doxorubicin.
Autophagy (2016-04-29)
Yang Wang, Kairong Shi, Li Zhang, Guanlian Hu, Jingyu Wan, Jiajing Tang, Sheng Yin, Jiandong Duan, Ming Qin, Neng Wang, Dandan Xie, Xinle Gao, Huile Gao, Zhirong Zhang, Qin He
ABSTRACT
Hydroxychloroquine (HCQ) inhibits autophagy and therefore can sensitize some cancer cells to chemotherapy, but the high doses required limit its clinical use. Here we show that loading HCQ into liposomes (HCQ/Lip) decorated with a pH-sensitive TH-RGD targeting peptide (HCQ/Lip-TR) can concentrate HCQ in B16F10 tumor cells and lysosomes. HCQ/Lip-TR was efficiently internalized as a result of its ability to bind ITGAV-ITGB3/integrin αvβ3 receptors highly expressed on the tumor cell surface and to undergo charge reversal from anionic at pH 7.4 to cationic at pH 6.5. Studies in vitro at pH 6.5 showed that the intracellular HCQ concentration was 35.68-fold higher, and lysosomal HCQ concentration 32.22-fold higher, after treating cultures with HCQ/Lip-TR than after treating them with free HCQ. The corresponding enhancements observed in mice bearing B16F10 tumors were 15.16-fold within tumor cells and 14.10-fold within lysosomes. HCQ/Lip-TR was associated with milder anemia and milder myosuppressive reductions in white blood cell and platelet counts than free HCQ, as well as less accumulation in the small intestine, which may reduce risk of intestinal side effects. In addition, co-delivering HCQ/Lip-TR with either free doxorubicin (DOX) or liposomal DOX improved the ability of DOX to inhibit tumor growth. Biochemical, electron microscopy and immunofluorescence experiments confirmed that HCQ/Lip-TR blocked autophagic flux in tumor cells. Our results suggest that loading HCQ into Lip-TR liposomes may increase the effective concentration of the inhibitor in tumor cells, allowing less toxic doses to be used.
Sigma-Aldrich
Lysosome Isolation Kit, sufficient for 25 g (tissue), sufficient for 20 mL (packed cells), enrichment of lysosomes from tissues and packed cells
Sigma-Aldrich
Mammalian Cell Lysis Kit