Merck
  • Silver nanoparticles for the detection of cell surface antigens in mass cytometry.
Silver nanoparticles for the detection of cell surface antigens in mass cytometry.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2016-06-29)
Axel Ronald Schulz, Silke Stanislawiak, Sabine Baumgart, Andreas Grützkau, Henrik E Mei
ABSTRACT
Mass cytometry has pioneered >40-parameter single-cell analyses that allow for the characterization of complex cellular networks at unprecedented depth. Up to 135 parameters can be simultaneously detected, but limited availability of metal tags suitable for labeling of specific probes prevents optimal exploitation of the analytical capacity of mass cytometers. To this end, we here establish the application of elemental silver nanoparticles (AgNP) of different size for reporting cell surface antigens on human leukocytes in mass cytometry assays. The mass channels at 107 Da and 109 Da are uniquely occupied by silver isotopes and do not interfere with other mass cytometry reagents. Streptavidin-coated AgNP (SA-AgNP) facilitated distinct and specific detection of various antigens, such as CD8, CD244 and CD294 on peripheral blood leukocytes pre-incubated with respective biotinylated primary antibodies. Signal intensities elicited by 40 nm-sized AgNP allowed specific detection of the low abundance antigen CD25 on both, peripheral blood regulatory T cells and CD25
Sigma-Aldrich
Rhodium(III) chloride, 98%
Pricing and availability is not currently available.