Merck
  • A TRPV1-to-secretagogin regulatory axis controls pancreatic β-cell survival by modulating protein turnover.
A TRPV1-to-secretagogin regulatory axis controls pancreatic β-cell survival by modulating protein turnover.
The EMBO journal (2017-06-24)
Katarzyna Malenczyk, Fatima Girach, Edit Szodorai, Petter Storm, Åsa Segerstolpe, Giuseppe Tortoriello, Robert Schnell, Jan Mulder, Roman A Romanov, Erzsébet Borók, Fabiana Piscitelli, Vincenzo Di Marzo, Gábor Szabó, Rickard Sandberg, Stefan Kubicek, Gert Lubec, Tomas Hökfelt, Ludwig Wagner, Leif Groop, Tibor Harkany
ABSTRACT
Ca
Sigma-Aldrich
MISSION® esiRNA, targeting human SCGN
Sigma-Aldrich
Arachidonylethanolamide, ≥97.0% (TLC), oil
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)