Merck
  • Enantioseparation of nornicotine in tobacco by ultraperformance convergence chromatography with tandem mass spectrometry.
Enantioseparation of nornicotine in tobacco by ultraperformance convergence chromatography with tandem mass spectrometry.
Journal of separation science (2017-09-30)
Huimin Deng, Ying Wang, Zhaoyang Bian, Shanshan Liu, Ziyan Fan, Zhonghao Li, Fei Yang, Gangling Tang
ABSTRACT
Nornicotine, an alkaloid constituent of tobacco, is a precursor to the carcinogen N-nitrosonornicotine that is produced during the curing and processing of tobacco. Accumulating evidence reveals that nornicotine enantiomers have different neurochemical and behavioral effects. In the present study, an accurate and rapid method was developed for the enantioseparation of (R)-(+)-nornicotine and (S)-(-)-nornicotine enantiomers in tobacco by ultra-performance convergence chromatography with tandem mass spectrometry. Chromatographic conditions were investigated to achieve the optimal resolution of two enantiomers. Results indicated that (R)-(+)-nornicotine and (S)-(-)-nornicotine could be separated within 5 min when ammonium hydroxide was added into the cosolvent, and the best resolution (R