Merck
  • Upregulated LAMB3 increases proliferation and metastasis in thyroid cancer.
Upregulated LAMB3 increases proliferation and metastasis in thyroid cancer.
OncoTargets and therapy (2018-01-11)
Yinghao Wang, Yixiang Jin, Adheesh Bhandari, Zhihan Yao, Fan Yang, Yiyuan Pan, Zhouci Zheng, Shixu Lv, Ouchen Wang
ABSTRACT
Thyroid cancer is the most commonly reported endocrine malignancy, and its increased incidence has been the highest in all human tumors in recent decades. To investigate the mechanism of papillary thyroid cancer (PTC) occurrence and progression, we performed RNA sequencing and found an upregulated gene, We analyzed We demonstrated that This study indicated that
Sigma-Aldrich
MISSION® esiRNA, targeting human LAMB3
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder