Merck
HPLC Analysis of Anions and Cations on SeQuant® ZIC-HILIC
HPLC Analysis of Anions and Cations on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
Sequant ZIC-HILIC, PEEK, 150 x 2.1 mm, 3.5 μm, 200 Å (1.50448.0001)
mobile phase
[A] acetonitrile; [B] 20 mM ammonium formate, pH 3
gradient
80% A held for 3 min; to 20% A in 7 min; held at 20% A for 3 min; to 80% A in 2 min; held at 80% A for 10 min
flow rate
0.3 mL/min
column temp.
40 °C
detector
ELSD (SEDEX 85LT, 40°C, 3.5 bar)
injection
2 μL
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany