Merck
HPLC Analysis of Substance P and Its Fragments on Discovery® C18
HPLC Analysis of Substance P and Its Fragments on Discovery® C18 application for HPLC
Materials
related product
Product No.
Description
Pricing
Discovery® C18 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] water + 0.1% TFA; [B] acetonitrile + 0.1% TFA
gradient
linear; 5-29%B (6 min); 29-34B (5 min)
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 220 nm
sample
Substance P in initial mobile phase
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany