Merck
Conformational Sampling of Small Molecules With iCon: Performance Assessment in Comparison With OMEGA.
Frontiers in chemistry (2018-07-05)
Giulio Poli, Thomas Seidel, Thierry Langer
RÉSUMÉ
Herein we present the algorithm and performance assessment of our newly developed conformer generator iCon that was implemented in LigandScout 4.0. Two data sets of high-quality X-ray structures of drug-like small molecules originating from the Protein Data Bank (200 ligands) and the Cambridge Structural Database (481 molecules) were used to validate iCon's performance in the reproduction of experimental conformations. OpenEye's conformer generator OMEGA was subjected to the same evaluation and served as a reference software in this analysis. We tested several setting patterns in order to identify the most suitable and efficient ones for conformational sampling with iCon; equivalent settings were also tested on OMEGA in order to compare the results obtained from the two programs and better assess iCon's performance. Overall, this study proved that iCon is able to generate reliable representative conformational ensembles of drug-like small molecules, yielding results comparable to those showed by OMEGA, and thus is ready to serve as a valuable tool for computer-aided drug design.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
N-Octanoyl-N-methylglucamine, ≥97% (GC)