Merck
  • Conformationally defined neurotransmitter analogues. Selective inhibition of glutamate uptake by one pyrrolidine-2,4-dicarboxylate diastereomer.
Conformationally defined neurotransmitter analogues. Selective inhibition of glutamate uptake by one pyrrolidine-2,4-dicarboxylate diastereomer.
Journal of medicinal chemistry (1991-02-01)
R J Bridges, M S Stanley, M W Anderson, C W Cotman, A R Chamberlin
RÉSUMÉ
In order to determine the conformational requirements for binding of L-glutamate to the proteins involved in the process of neurotransmission, rigid analogues containing an embedded glutamate moiety have been prepared. These "conformer mimics", the pyrrolidine-2,4-dicarboxylates 4, 7, 11, and 14, were synthesized from commercially available trans-4-hydroxy-L-proline and cis-4-hydroxy-D-proline, and then were tested for their ability to inhibit the high-affinity transport of [3H]-L-glutamate into synaptosomes and to block the binding of radioligands to the NMDA (N-methyl-D-aspartate), KA (kainate), and QA (quisqualate) glutamate neurotransmitter receptor sites. While none of the four analogues binds effectively to the excitatory receptors, the L-trans-isomer 7 is a potent and selective competitive inhibitor of L-glutamate transport. These results delineate a specific structural/conformational preference for binding to the uptake system that is distinct from that required for binding to the NMDA, KA, and QA receptors.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
L-trans-Pyrrolidine-2,4-dicarboxylic acid, ≥98%