Merck
Propofol increases agonist efficacy at the GABA(A) receptor.
Brain research (2000-03-11)
S M O'Shea, L C Wong, N L Harrison
RÉSUMÉ
Using the whole-cell patch-clamp technique, we have determined that propofol, but not midazolam, increases the efficacy of piperidine-4-sulphonic acid (P4S), a partial agonist at alpha1beta1gamma2s, GABA(A) receptors expressed in HEK 293 cells. These findings are consistent with the idea that propofol facilitates receptor gating, while midazolam increases receptor occupancy by the agonist.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Piperidine-4-sulfonic acid