Merck
Biodistribution, pharmacokinetics and toxicology of Ag2S near-infrared quantum dots in mice.
Biomaterials (2013-02-19)
Yan Zhang, Yejun Zhang, Guosong Hong, Wei He, Kun Zhou, Kai Yang, Feng Li, Guangcun Chen, Zhuang Liu, Hongjie Dai, Qiangbin Wang
RÉSUMÉ
Ag2S quantum dots (QDs) have been demonstrated as a promising near-infrared II (NIR-II, 1.0-1.4 μm) emitting nanoprobe for in vivo imaging and detection. In this work, we carefully study the long-term in vivo biodistribution of Ag2S QDs functionalized with polyethylene glycol (PEG) and systematically examine the potential toxicity of Ag2S QDs over time. Our results show that PEGylated-Ag2S QDs are mainly accumulated in the reticuloendothelial system (RES) including liver and spleen after intravenous administration and can be gradually cleared, mostly by fecal excretion. PEGylated-Ag2S QDs do not cause appreciable toxicity at our tested doses (15 and 30 mg/kg) to the treated mice over a period of 2 months as evidenced by blood biochemistry, hematological analysis and histological examinations. Our work lays a solid foundation for further biomedical applications of Ag2S QDs as an important in vivo imaging agent in the NIR-II region.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Silver sulfide, 99.9% trace metals basis