Merck
Effect of caffeine on G2 repair and its reversion by adenosine.
Mutation research (1982-12-01)
A González-Fernández, J F López-Sáez
RÉSUMÉ
Chromosome damage induced in root meristems of Allium cepa L. by an 18-h treatment with 5-aminouracil (AU) was enhanced by 2-h pulses with 5 mM caffeine, the most effective pulse being given from the 8th to the 10th h after AU. Chromosome damage was detected by the percentage of anaphases or telophases showing chromosomal aberrations. This caffeine effect was partially reversed by adenosine. We suggest that this caffeine anti-repair effect could be due to a competition between methylxanthine and an adenine-nucleotide derivative (ATP?) required for some step(s) of G2-prophase repair.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
5-Aminouracil, 98%