Merck
Synthesis of azomethine imines using an intramolecular alkyne hydrohydrazination approach.
The Journal of organic chemistry (2013-07-31)
Ashley D Hunt, Isabelle Dion, Nicolas Das Neves, Sandrine Taing, André M Beauchemin
RÉSUMÉ
Azomethine imines can be accessed upon heating appropriate alkynylhydrazide precursors. This simple thermal hydroamination approach allows the formation of five- and six-membered dipoles in modest to excellent yields. The structure of the acyl group is important to minimize side reactions and allow the isolation of the azomethine imines by column chromatography.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Hydralazine hydrochloride