Skip to Content
Merck

ATE514317841

AstaTech, Inc.

1-METHYL-PIPERAZINE-2-CARBOXYLIC ACID

Synonym(s):

1-methyl-2-piperazinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H12O2N2
Molecular Weight:
144.17
MDL number:
Product ATE514317841 is not currently sold in your country.


Inchi Code

1S/C6H12N2O2/c1-8-3-2-7-4-5(8)6(9)10/h5,7H,2-4H2,1H3,(H,9,10)

Inchi Key

CCOAGJXNPXLMGI-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library