Skip to Content
Merck

BDPH9BCCE1C8

Bide Pharmatech

2-Chloro-4,6-dimethoxy-1,3,5-triazine

Synonym(s):

2-chloro-4,6-dimethoxy-1,3,5-triazine

Select a Size

Change View

About This Item

Linear Formula:
C5H6O2N3Cl1
CAS Number:
Molecular Weight:
175.57
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H6ClN3O2/c1-10-4-7-3(6)8-5(9-4)11-2/h1-2H3

Inchi Key

GPIQOFWTZXXOOV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder

Melting Point

71-74C



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library